Anti-PBP Polyclonal Antibody
BSBTA01668
- Antibody type:Primary
- Antigen symbol:PBP
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for PBP detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: PBP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat