Anti-Kininogen-1/KNG1 Polyclonal Antibody
Catalog # BSBTPB9278
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Kininogen-1/KNG1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for KININOGEN 1/KNG1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. KININOGEN 1/KNG1 information: Molecular Weight: 71957 MW; Subcellular Localization: Secreted, extracellular space; Tissue Specificity: Secreted in plasma. T-kinin is detected in malignant ovarian, colon and breast carcinomas, but not in benign tumors.
Type: Primary
Antigen: Kininogen-1/KNG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human