Anti-HMOX2/Heme Oxygenase 2 Polyclonal Antibody
BSBTPB9213
- Antibody type:Primary
- Antigen name:Heme Oxygenase 2
- Antigen symbol:HMOX2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for HEME OXYGENASE 2/HMOX2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. HEME OXYGENASE 2/HMOX2 information: Molecular Weight: 36033 MW; Subcellular Localization: Microsome. Endoplasmic reticulum.
Type: Primary
Antigen: HMOX2/Heme Oxygenase 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human