Specifications
- Antibody type:Primary
- Antigen symbol:HEF1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for HEF1/NEDD9 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. HEF1/NEDD9 information: Molecular Weight: 92861 MW; Subcellular Localization: Cytoplasm, cell cortex. Nucleus. Golgi apparatus. Cell projection, lamellipodium. Cytoplasm. Cell junction, focal adhesion. Localizes to both the cell nucleus and the cell periphery and is differently localized in fibroblasts and epithelial cells. In fibroblasts is predominantly nuclear and in some cells is present in the Golgi apparatus. In epithelial cells localized predominantly in the cell periphery with particular concentration in lamellipodia but is also found in the nucleus. Isoforms p105 and p115 are predominantly cytoplasmic and associate with focal adhesions while p55 associates with mitotic spindle; Tissue Specificity: Widely expressed. Higher levels detected in kidney, lung, and placenta. Also detected in T-cells, B-cells and diverse cell lines. The protein has been detected in lymphocytes, in diverse cell lines, and in lung tissues.
Type: Primary
Antigen: HEF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human