Anti-Galectin 10 Polyclonal Antibody
Catalog # BSBTA01350
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Galectin 10
- Antigen symbol:LGALS10
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Galectin 10 detection. Tested with WB, Direct ELISA in Human.
Type: Primary
Antigen: Galectin 10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human