Order Entry
Italy
ContactUsLinkComponent
Anti-CTNNA1 Polyclonal Antibody
Anti-CTNNA1 Polyclonal Antibody
Catalog # BSBTPB9137
Supplier:  Boster Bio
undefined
Anti-CTNNA1 Polyclonal Antibody
Catalog # BSBTPB9137
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Catenin Alpha 1
  • Antigen symbol:
    CTNNA1
  • Clonality:
    Polyclonal
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for ALPHA 1 CATENIN/CTNNA1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. ALPHA 1 CATENIN/CTNNA1 information: Molecular Weight: 100071 MW; Subcellular Localization: Isoform 1: Cytoplasm, cytoskeleton. Cell junction, adherens junction. Cell membrane; Peripheral membrane protein; Cytoplasmic side. Cell junction. Found at cell-cell boundaries and probably at cell-matrix boundaries; Tissue Specificity: Expressed ubiquitously in normal tissues.

Type: Primary
Antigen: CTNNA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat