Order Entry
Italy
ContactUsLinkComponent
Anti-complement C5a Polyclonal Antibody
Anti-complement C5a Polyclonal Antibody
Catalog # BSBTPB9187
Supplier:  Boster Bio
undefined
Anti-complement C5a Polyclonal Antibody
Catalog # BSBTPB9187
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    complement C5a
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for C5A/C5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Rat. C5A/C5 information: Molecular Weight: 8981 MW; Subcellular Localization: Secreted.

Type: Primary
Antigen: complement C5a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Rat