Anti-CNTF Polyclonal Antibody
BSBTPB9067
- Antibody type:Primary
- Antigen symbol:CNTF
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for CNTF detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Mouse. CNTF information: Molecular Weight: 22854 MW; Subcellular Localization: Cytoplasm; Tissue Specificity: Nervous system.
Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat