Anti-CD23/FCER2 Polyclonal Antibody
Catalog # BSBTPB9051
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CD23/FCER2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for CD23/FCER2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CD23/FCER2 information: Molecular Weight: 37648 MW; Subcellular Localization: Cell membrane ; Single-pass type II membrane protein . Cell membrane ; Lipid-anchor .
Type: Primary
Antigen: CD23/FCER2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat