Anti-BAFF Receptor Polyclonal Antibody
Catalog # BSBTA02865-2
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:BAFF Receptor
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for BAFF Receptor detection. Tested with WB, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: BAFF Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat