Anti-AP2 Alpha Polyclonal Antibody
BSBTPB9118
- Antibody type:Primary
- Antigen symbol:AP2 Alpha
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for AP2 ALPHA/TFAP2A detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. AP2 ALPHA/TFAP2A information: Molecular Weight: 48062 MW; Subcellular Localization: Nucleus .
Type: Primary
Antigen: AP2 Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat