EDTA disodium salt dihydrate ≥99%, white crystalline powder for molecular biology
:6381-92-6
:
:C₁₀H₁₄N₂Na₂O₈·2H₂O
:MFCD00150037
:372.24 g/mol
:238…248 °C
:Ambient
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- pH (1% aq soln):4-6
- Loss on Drying:≤10%
Frequently Bought Together