Order Entry
Italy
ContactUsLinkComponent
L(+)-Histidine ≥99%, white crystalline powder cell culture reagent
L(+)-Histidine ≥99%, white crystalline powder cell culture reagent
Supplier:  MP Biomedicals
Certificates
undefined
L(+)-Histidine ≥99%, white crystalline powder cell culture reagent
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • pH (1% aq soln):
    7.0-8.5
  • Loss on Drying:
    ≤0.5%
  • Optical Rotation (c=10, 4N HCl):
    +11.0° to +13.5°

Specification Test Results

Frequently Bought Together