
Proguanil hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:637-32-1
:
Formula:C₁₁H₁₆ClN₅·HCl
MDL Number:MFCD01732193
:290.20 g/mol
Melting Pt:245…250 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C)
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together