1,8-Diazafluoren-9-one
:54078-29-4
:
:C₁₁H₆N₂O
:MFCD00082335
:182.18 g/mol
:229…233 °C
:GHS07
:Refrigerator
- Appearance:Yellow to orange Powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 %
- Dye content:≥70.0 %
Frequently Bought Together