Entecavir monohydrate 99%
Glass bottle, 1 g
Catalog # ACRO461050010
Supplier: Thermo Fisher Scientific
Entecavir monohydrate 99%
Catalog # ACRO461050010
Supplier: Thermo Fisher Scientific
CAS Number: 209216-23-9
Jump to:
Product Details & Documents
Synonyms: 2-amino-9-((1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl)-1,9-dihydro-6H-purin-6-one hydrate, 9-[(1S,3R,4S)-4-Hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]guanine monohydrate
- CAS Number:209216-23-9
- Formula:C₁₂H₁₇N₅O₄
- MDL Number:MFCD09754448
- Molecular Weight:295.29 g/mol
- Melting Pt:259 °C (dec.)
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:5.8 to 7 %
- HPLC:≥98.5 %
- Optical rotation:+24° to +28° (1 % in DMF/methanol)
Specifications
- Pack type:Glass bottle
- Pk:1 g



