Order Entry
Italy
ContactUsLinkComponent
2-Methyl-4-isothiazolin-3-one 50% in aqueous solution
Corrosive
Toxic
Environment
2-Methyl-4-isothiazolin-3-one 50% in aqueous solution
Certificates
undefined
Corrosive
Toxic
Environment
2-Methyl-4-isothiazolin-3-one 50% in aqueous solution

Specification Test Results

  • Appearance:
    Clear colorless to yellow Solution
  • pH:
    5.0 to 7.0 (1 % in water)
  • Concentration:
    ≥50 %

Specification Test Results

Frequently Bought Together