
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15163-36-7
:
Formula:C₁₅H₃₃NO₃S
MDL Number:MFCD00036908
:307.4924 g/mol
Melting Pt:229 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:227 to 233 °C
- Loss on drying:≤2 %
- Solubility:(10% in water) Clear colorless solution
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together