Order Entry
Italy
ContactUsLinkComponent
4-Iodo-1-tritylimidazole 98%
Irritant
4-Iodo-1-tritylimidazole 98%
Certificates
undefined
Irritant
4-Iodo-1-tritylimidazole 98%

Specification Test Results

  • Appearance:
    White to off-white Powder or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    222 to 229 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Frequently Bought Together