Aluminium potassium sulphate, anhydrous, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10043-67-1
:
Formula:AlKO₈S₂
:258.21 g/mol
Boiling Pt:200 °C (760 mmHg)
Melting Pt:92 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:≥90 %
- Loss on drying:≤1 %
- Ammonium (NH4):≤0.3 %
- Iron (Fe):≤0.02 %
- Heavy metals (as Pb):≤0.003 %
Specification Test Results
Frequently Bought Together