Order Entry
Italy
Orders LinkContactUsLinkComponent
Finasteride 98%

Finasteride 98%

Irritant
Health
Finasteride 98%
Certificates
Finasteride 98%

2 Options Available Below

Product Details & Documents
Synonyms:  N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
  • CAS Number:98319-26-7
  • Formula:C₂₃H₃₆N₂O₂
  • MDL Number:MFCD00869737
  • Molecular Weight:372.55 g/mol
  • Melting Pt:252…254 °C
  • Storage Temperature:Freezer
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.5 %
  • HPLC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 162,00 €
    162,00 €
    1 * 1 g
       162,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 495,00 €
    495,00 €
    1 * 5 g
       495,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.