Order Entry
Italy
ContactUsLinkComponent
2-(Isocyanatomethyl)furan 97%
Flammable
Irritant
Health
2-(Isocyanatomethyl)furan 97%
Certificates
undefined
Flammable
Irritant
Health
2-(Isocyanatomethyl)furan 97%

Specification Test Results

  • Appearance:
    Clear colorless to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4750 to 1.4770 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Frequently Bought Together