Order Entry
Italy
Orders LinkContactUsLinkComponent
Capecitabine 99%

Capecitabine 99%

Glass bottle, 100 mg
Catalog # ACRO456241000
Irritant
Health
Capecitabine 99%
Catalog # ACRO456241000
CAS Number:  154361-50-9
Capecitabine 99%
Capecitabine 99%

Glass bottle, 100 mg

CAS Number:  154361-50-9
Price
237,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  5'-Deoxy-5-fluoro-N4-[(pentyloxy)carbonyl]cytidine, Pentyl (1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl)carbamate
  • CAS Number:154361-50-9
  • Formula:C₁₅H₂₂FN₃O₆
  • MDL Number:MFCD00930626
  • Molecular Weight:359.35 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 %
  • HPLC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg

Specifications