
7-Azaindole-5-boronic acid pinacol ester 97%
Catalog # ACRO455850010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light brown Powder
- Infrared spectrum:Conforms
- Melting point:243 to 249 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
Melting Pt:243…245.5 °C
:244.101 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:754214-56-7
Formula:C₁₃H₁₇BN₂O₂
MDL Number:MFCD08060937