
4-tert-Butylcyclohexylamine cis- and trans-mixture 97%
:5400-88-4
:
:C₁₀H₂₁N
:MFCD00013763
:155.28 g/mol
:0.87 g/cm³
:79 °C
:Ambient
:2735
:8,III
:GHS05
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4630 to 1.4660 (20°C, 589 nm)
- GC:≥96.0 % (total of isomers)
Frequently Bought Together