Order Entry
Italy
Orders LinkContactUsLinkComponent
Chloroquine diphosphate salt 98%

Chloroquine diphosphate salt 98%

Irritant
Chloroquine diphosphate salt 98%
Certificates
Chloroquine diphosphate salt 98%

3 Options Available Below

Product Details & Documents
Synonyms:  N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate
  • CAS Number:50-63-5
  • Formula:C₁₈H₂₆ClN₃
  • MDL Number:MFCD00069852
  • Molecular Weight:515.86 g/mol
  • Melting Pt:200 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 % (105°C, 16 h)
  • HPLC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 52,00 €
    52,00 €
    1 * 25 g
       52,00 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 168,00 €
    168,00 €
    1 * 100 g
       168,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 395,00 €
    395,00 €
    1 * 500 g
       395,00 €