Order Entry
Italy
ContactUsLinkComponent
4-Acetamidobenzenesulphonyl azide 97%
Irritant
4-Acetamidobenzenesulphonyl azide 97%
Certificates
undefined
Irritant
4-Acetamidobenzenesulphonyl azide 97%

Specification Test Results

  • Appearance:
    White to grey to brown Crystalline powder and or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    107 to 111 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Frequently Bought Together