
L-Dehydroascorbic acid 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:490-83-5
:
Formula:C₆H₆O₆
MDL Number:MFCD00066467
:174.108 g/mol
Melting Pt:228 °C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to light brown Powder
- Infrared spectrum:Conforms
- HPLC:≥95.0 % (ELSD)
Specification Test Results
Frequently Bought Together