Order Entry
Italy
ContactUsLinkComponent
 
 
Propyl-3,4,5-trihydroxybenzoate 97.0-103.0% (by UV method) Ph. Eur., pure
  APPC141962.1211
 :  
 
Propyl-3,4,5-trihydroxybenzoate 97.0-103.0% (by UV method) Ph. Eur., pure
  APPC141962.1211
 :  

 

  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay (UV, dried substance):
    97,0 - 103,0%
  • Sulphated ash:
    Max. 0,1%
  • Loss on drying (105 °C):
    Max. 0,5%
  • Identity:
    Passes test
  • Appearance of solution:
    Passes test
  • Chloride:
    Max. 0,01%
  • Total chlorine:
    Max. 0,02%
  • Gallic acid (TLC):
    Passes test
  • Zinc (Zn):
    max. 0,0025%
  • Form:
    Solid

 

 

 :
 :212.2 g/mol
 :GHS07
 :Ambient
 :121-79-9
 :C₁₀H₁₂O₅
 :MFCD00002196