Order Entry
Italy
Orders LinkContactUsLinkComponent
Enrofloxacin, Reference Standards

Enrofloxacin, Reference Standards

Supplier:   USP
Enrofloxacin, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1235900.
  • 1235911.
  • Description:
  • Enrofloxacin
  • Enrofloxacin Related Compound Mixture
  • CAS No.:
  • 93106-60-6
  • Formula:
  • C₁₉H₂₂FN₃O₃
  • UN No.:
  • 3077
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1235900.
    Enrofloxacin
    Enrofloxacin
    USP reference standard
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 393,00 €
    393,00 €
    1 * 200 mg
       393,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1235900.
    Description
    Enrofloxacin
    CAS No.
    93106-60-6
    Formula
    C₁₉H₂₂FN₃O₃
    UN No.
    3077
  • 1235911.
    Enrofloxacin Related Compound Mixture
    Enrofloxacin Related Compound Mixture
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 310,00 €
    1 310,00 €
    1 * 50 mg
       1 310,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1235911.
    Description
    Enrofloxacin Related Compound Mixture
    CAS No.
    Formula
    UN No.
    3077