Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Poloxamer 407, Ph.Eur., RS
Organic reference standard, Poloxamer 407, Ph.Eur., RS
Catalog # EDQMP2164030
Supplier:  EDQM
CAS Number:  
Organic reference standard, Poloxamer 407, Ph.Eur., RS
Catalog # EDQMP2164030
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    9003-11-6
  • Description:
    Poloxamer 407
  • Formula:
    C₅H₁₀O₂
  • Description:
    Poloxamer 407

Specifications

About this item

  • In Monograph 1464
  • Solubility in water and ethanol at 20 °C