Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Biological activity:Bioactivity was measured in a cell proliferation assay using anti-CD3 activated human peripheral mononuclear cells.
- Protein synonyms:Interleukin-27
- Protein/peptide name:IL-27 non-lytic
- Purity:>98% (SDS-PAGE)
- Sequence:26-243
- Formulation:Lyophilized from 0.2um-filtered solution in PBS.
Specifications
About this item
Interleukin-27 (IL-27) is a heterodimeric group 2 receptor ligand molecule that belongs to the IL-6/IL-12 family of long type I cytokines. It is composed of EBI3 (EBV-induced gene 3), a 34 kDa glycoprotein that is related to the p40 subunit of IL-12 and IL-23, and p28, the cloned 28 kDa glycoprotein that is related to the p35 chain of IL-12. IL-27 is expressed by monocytes, endothelial cells and dendritic cells. IL-27 binds to and signals through a heterodimeric receptor complex composed of WSX1 (TCCR) and gp130. Evidence suggests IL-27 interacts only with WSX-1. IL-27 has both anti- and proinflammatory properties. As an antiinflammatory, IL-27 seems to induce a general negative feedback program that limits T and NK-T cell activity. At the onset of infection, IL-27 induces an IL-12 receptor on naie CD4+ T cells, making them susceptible to subsequent IL-12 activity (and possible Th1 development). Notably, IL-12 family cytokines are both induced and inhibited by bacterial products. Microbes promote IL-27 secretion through TLR4, and also block IL-27 production via C5a induction.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.