Order Entry
Italy
ContactUsLinkComponent
Mouse recombinant IL12
Mouse recombinant IL12
Catalog # PRSI92-631
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant IL12
Catalog # PRSI92-631
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-12|NK cell|Interleukin-12 subunit alpha|Cytotoxic lymphocyte maturation factor 35 kDa subunit|IL-12A|IL-12 subunit p35|Interleukin 12|CLMF p35
  • Protein/peptide name:
    IL12
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    57.5 kD
  • Sequence:
    Met23-Ser335&Arg23-Ala215
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

IL-12 is involved in the differentiation of naive T cells into Th1 cells. It is known as a T cell-stimulating factor, which can stimulate the growth and function of T cells. It stimulates the production of interferon-gamma (IFN- gamma ) and tumor necrosis factor-alpha (TNF- alpha) from T cells and natural killer (NK) cells, and reduces IL-4 mediated suppression of IFN- gamma . T cells that produce IL-12 have a coreceptor, CD30, which is associated with IL-12 activity. IL-12 plays an important role in the activities of natural killer cells and T lymphocytes. IL-12 mediates enhancement of the cytotoxic activity of NK cells and CD8+ cytotoxic T lymphocytes.

This recombinant protein can be used for biological assays. For research use only.