Order Entry
Italy
ContactUsLinkComponent
Human recombinant HOXB4 (from E. coli)
Human recombinant HOXB4 (from E. coli)
Catalog # PRSI92-525
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant HOXB4 (from E. coli)
Catalog # PRSI92-525
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    HOXB4|Homeobox protein Hox-2F|HOX-2F|Homeobox protein Hox-B4|HOX2F|Homeobox protein Hox-2.6
  • Protein/peptide name:
    HOXB4
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    29.8 kD
  • Sequence:
    Met1-Leu251
  • Formulation:
    Supplied as a 0.2 um filtered solution of 4mM HCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Homeobox B4 (HOXB4) is encoded by the HOXB4 gene which is a member of the the class I homeobox (HOX) gene family and encodes a nuclear protein with a homeobox DNA-binding domain. These genes are master control regulators of developmental programs including embryonic and adult hematopoiesis. Multiple HOX genes, including HOXB4, are highly expressed in the hematopoietic stem cells (HSC) compartment. HOXB4 gene can act in opposite ways when expressed by different cells, promoting the proliferation of stem cells whilst activating the apoptotic pathway in some embryonic structures. The protein HOXB4, as a homeodomain transcription factor, has been shown to be an important regulator of stem cell renewal and hematopoiesis. Incellular or ectopic expression of HOXB4 expands hematopoietic stem and progenitor cells in vivo and in vitro, making it a potential candidate for therapeutic stem cell expansion.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.