Mouse recombinant HLADG
Catalog # PRSI92-450
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-mFc-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Cluster of Differentiation 74|HLA class II histocompatibility antigen gamma chain|CD74|HLA-DR antigens-associated invariant chain
- Protein/peptide name:HLADG
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:44.5 kD
- Sequence:Gln56-Leu215
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Mouse HLA class II histocompatibility antigen gamma chain (CD74), is a single-pass type II membrane protein that in humans is encoded by the CD74 gene. It contains 1 thyroglobulin type-1 domain. CD74 Plays a critical role in MHC class II antigen processing by stabilizing peptide-free class II alpha/beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to compartments where peptide loading of class II takes place.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-mFc-6 His tag
This recombinant protein can be used for biological assays. For research use only.