Human recombinant Annexin A3 (from E. coli)
Catalog # PRSI92-273
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:ANXA3|Placental anticoagulant protein III|ANX3|Lipocortin III|Annexin III|35-alpha calcimedin|PAP-III|2-cyclic phosphate 2-phosphohydrolase|Annexin-3|Inositol 1
- Protein/peptide name:Annexin A3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:36.4 kD
- Sequence:Met1-Asp323
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Annexin A3(ANXA3)contains 4 annexin repeats and belongs to the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. It is abnormally expressed in fetuses of both IVF and ICSI, which may contribute to the increase risk of birth defects in these ART. ANXA3 is Inhibitor of phospholipase A2 and cleaves the cyclic bond of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. it also also play a role in anti-coagulant properties.
This recombinant protein can be used for biological assays. For research use only.