Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant PDCD10 (from E. coli)
  PRSI92-082
 :  
Human recombinant PDCD10 (from E. coli)
  PRSI92-082
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    TF-1 Cell Apoptosis-Related Protein 15|TFAR15|PDCD10|CCM3|Programmed Cell Death Protein 10|Cerebral Cavernous Malformations 3 Protein
  • Protein/peptide name:
    PDCD10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    24.9 kD
  • Sequence:
    Met1-Ala212
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 25mM TrisHCl,pH7.3. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays

 

 

Programmed Cell Death Protein 10 (PDCD10) belongs to the PDCD10 family. PDCD10 exists as a homodimer and is widely expressed. PDCD10 can increase mitogen-activated protein kinase activity and MST4 activity. PDCD10 is required for normal cardiovascular development and normal angiogenesis, vasculogenesis and hematopoiesis during embryonic development. Defects in PDCD10 are the cause of cerebral cavernous malformations type 3.

  • This recombinant protein can be used for biological assays. For research use only.