Order Entry
Italy
ContactUsLinkComponent
Mouse recombinant TWEAK receptor
Mouse recombinant TWEAK receptor
Catalog # PRSI91-946
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant TWEAK receptor
Catalog # PRSI91-946
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Tweak-receptor|TWEAK R|Fibroblast growth factor-inducible immediate-early response protein 14|TweakR|Fibroblast growth factor-regulated protein 2|TNFRSF12A|TNFRSF12|Tumor necrosis factor receptor superfamily member 12A
  • Protein/peptide name:
    TWEAK Receptor
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    32.6 kD
  • Sequence:
    Glu28-Trp79
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Tumor necrosis factor receptor superfamily member 12A(Tnfrsf12a) is a single-pass type I membrane protein and contains 1 TNFR-Cys repeat. It is weak inducer of apoptosis in some cell types.It promotes angiogenesis and it is the proliferation of endothelial cells. It may modulate cellular adhesion to matrix proteins.TNFR binds specifically to tumor necrosis factor (TNF) and blocks its interaction with cell surface TNF receptors. TNF is a naturally occurring cytokine that is involved in normal inflammatory and immune responses. It plays an important role in the inflammatory processes of rheumatoid arthritis (RA), polyarticular-course juvenile rheumatoid arthritis (JRA), and ankylosing spondylitis and the resulting joint pathology. In addition, TNF plays a role in the inflammatory process of plaque psoriasis. Elevated levels of TNF are found in involved tissues and fluids of patients with RA, psoriatic arthritis, ankylosing spondylitis (AS), and plaque psoriasis. Two distinct receptors for TNF (TNFRs), a 55 kilodalton protein (p55) and a 75 kilodalton protein (p75), exist naturally as monomeric molecules on cell surfaces and in soluble forms. Biological activity of TNF is dependent upon binding to either cell surface TNFR.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-Fc tag