Order Entry
Italy
ContactUsLinkComponent
Mouse recombinant ephrin B2
Mouse recombinant ephrin B2
Catalog # PRSI91-941
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant ephrin B2
Catalog # PRSI91-941
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-Fc-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    EFNB2|Epl5|ELF-2|Lerk5.|Htkl|Elf2|Ephrin-B2|EPH-related receptor tyrosine kinase ligand 5|HTK ligand|Eplg5
  • Protein/peptide name:
    Ephrin B2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    49.6 kD
  • Sequence:
    Arg29-Glu227
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays

Specifications

About this item

Ephrin-B2 is a single-pass type I membrane protein and it contains 1 ephrin RBD (ephrin receptor-binding) domain. Ephrin-B2 belongs to the ephrin (EPH) family and it is cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. The ephrins and EPH-related receptors contain the largest subfamily of receptor protein-tyrosine kinases and have been associated with mediating developmental events, particularly in the nervous system and in erythropoiesis. Based upon their structures and sequence relationships, ephrins are allocated into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. It also binds to receptor tyrosine kinase including EPHA4, EPHA3 and EPHB4 and together with EPHB4 plays a central role in heart morphogenesis and angiogenesis through regulation of cell adhesion and cell migration.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-Fc-6 His tag