Order Entry
Italy
ContactUsLinkComponent
Human recombinant protocadherin 10 (from cells)
Human recombinant protocadherin 10 (from cells)
Catalog # PRSI91-934
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant protocadherin 10 (from cells)
Catalog # PRSI91-934
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    PCDH10|KIAA1400|Protocadherin-10
  • Protein/peptide name:
    Protocadherin 10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    102.4 kD
  • Sequence:
    Gln19-Thr715
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Protocadherin-10 (PCDH10) is a single-pass type I membrane protein that contains six extracellular cadherin domains, one transmembrane domain and one cytoplasmic tail differing from those of the classical cadherins. As a potential calcium-dependent cell-adhesion neuronal receptor, it may plays a role in the establishment and function of specific cell-cell connections in the brain. PCDH10 moderately expressed in all regions of the brain examined, as well as in testis and ovary, and low expression in all other tested tissues.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-Fc tag