Human recombinant Chromogranin-C (from cells)
PRSI91-860
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Chromogranin-C|SgII.|Secretogranin-2|Secretogranin II|CHGC|SCG2
- Protein/peptide name:Chromogranin-C
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:68.8 kD
- Sequence:Ser29-Met617
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Secretogranin-2 is also known as Chromogranin-C,Secretogranin II, SgII. In humans, it is encoded by the SCG2 gene.It belongs to the chromogranin/secretogranin protein family. Secretogranin-2 is a neuroendocrine secretory granule protein, which is the precursor for biologically active peptides. It derived secretoneurin was distributed with strong immunoreactivity in the somata of pelvic ganglion neurons,72% of which also contained tyrosine hydroxyldse,as well as in nerve terminals in the muscular layer and the lamina propria of the vas deferens.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag