Order Entry
Italy
ContactUsLinkComponent
Human recombinant IL-18 binding (from cells)
Human recombinant IL-18 binding (from cells)
Catalog # PRSI91-814
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL-18 binding (from cells)
Catalog # PRSI91-814
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Interleukin-18-binding protein|IL-18BP|Tadekinig-alfa and IL18BP
  • Protein/peptide name:
    IL-18 binding
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    44.7 kD
  • Sequence:
    Thr31-Gly194
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Interleukin-18-binding protein, also known as Tadekinig-alfa and IL18BP, contains 1 Ig-like C2-type domain. IL18BP is constitutively expressed and secreted in mononuclear cells. IL18BP functions as an IL18 inhibitor. IL18BP binds to IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production. It has been shown that IL18BP may be a promising molecular approach to inhibit neointimal hyperplasia and arteriosclerosis progression following coronary and peripheral angioplasty.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-Fc tag