Order Entry
Italy
ContactUsLinkComponent
Human recombinant ER Resident 27 (from cells)
Human recombinant ER Resident 27 (from cells)
Catalog # PRSI91-767
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ER Resident 27 (from cells)
Catalog # PRSI91-767
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Endoplasmic Reticulum Resident Protein 27|ERp27|ER Protein 27|C12orf46
  • Protein/peptide name:
    ER Resident 27
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.79 kD
  • Sequence:
    Glu26-Leu273
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Endoplasmic reticulum resident protein 27, also known as ER protein 27, C12orf46 and ERP27, is an endoplasmic reticulum luminal protein which is a member of the protein disulfide isomerase family. ERP27 contains one thioredoxin domain and does not contain a CXXC active site motif. ERP27 is widely expressed in many tissues; it has highest expression in pancreas, with lower levels in spleen, lung, kidney, thymus, and bone marrow. ERP27 interacts with PDIA3 and binds somatostatin-14 via hydrophobic interactions. ERP27 may act as a protease, protein disulfide isomerase, thiol-disulfide oxidase or phospholipase.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag