Order Entry
Italy
ContactUsLinkComponent
Human recombinant Galectin 3 (from cells)
Human recombinant Galectin 3 (from cells)
Catalog # PRSI91-660
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Galectin 3 (from cells)
Catalog # PRSI91-660
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    L-31|Laminin-binding protein|Galectin-3|IgE-binding protein|LGALS3|GALBP|CBP 35|Lectin L-29|Mac-2 antigen|35 kDa lectin|Gal-3|MAC2
  • Protein/peptide name:
    Galectin 3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    27.2 kD
  • Sequence:
    Ala2-Ile250
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4, 3mM DTT. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Galectin-3(LGALS3) is also known as Galactose-specific lectin 3, Mac-2 antigen, Carbohydrate-binding protein 35, Laminin-binding protein and Galactoside-binding protein. LGALS3 is highly expressed in early stages of papillary carcinoma, and lowly during tumor progression. LGALS3 is probably forms homo- or heterodimers and secreted by a non-classical secretory pathway and associates with the cell surface. LGALS3 plays an important role during the acquisition of vasculogenic mimicry and angiogenic properties. LGLAS3 takes part in an immune regulator to inhibit T-cell immune responses and promote tumor growth, as a result providing a new mechanism for tumor immune tolerance.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag