Mouse recombinant CXCL1
Catalog # PRSI91-628
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Mgsa|Growth-regulated alpha protein|Gro|Cxcl1|Scyb1|Platelet-derived growth factor-inducible protein KC|GRO alpha|Secretory protein N51|Gro1|C-X-C motif chemokine 1
- Protein/peptide name:CXCL1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:9.4 kD
- Sequence:Arg20-Lys96
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Cxcl1, also called Gro, Gro1, Mgsa or Scyb1, is short for growth-regulated alpha protein. The protein belongs to the intercrine alpha (chemokine CxC) family. The N-terminal processed form KC(5-72) of the protein is produced by proteolytic cleavage after secretion from bone marrow stromal cells, and shows a highly enhanced hematopoietic activity. Cxcl1 has chemotactic activity for neutrophils, and contributes to neutrophil activation during inflammation. Hematoregulatory chemokine, in vitro, suppresses hematopoietic progenitor cell proliferation.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag