Order Entry
Italy
ContactUsLinkComponent
Human recombinant S100 calcium binding A12 (from E. coli)
Human recombinant S100 calcium binding A12 (from E. coli)
Catalog # PRSI91-588
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant S100 calcium binding A12 (from E. coli)
Catalog # PRSI91-588
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    S100 calcium-binding protein A12|EN-RAGE|CAGC|Calcitermin|S100A12|Protein S100-A12|CGRP|Extracellular newly identified RAGE-binding protein|MRP-6|Migration inhibitory factor-related protein 6|Calgranulin-C|Calcium-binding protein in amniotic fluid 1|Neutrophil S100 protein|CAAF1
  • Protein/peptide name:
    S100 calcium binding A12
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10.6 kD
  • Sequence:
    Met1-Glu92
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Human EN-RAGE belongs to the S100/calgranulin superfamily. There are at least 21 different S100 proteins and the protein is 100% soluble in ammonium sulfate at neutral pH. S100 proteins play a role in regulation of protein phosphorylation, transcription factors, the dynamics of cytoskeleton constituents, enzyme activities, cell growth and differentiation, and the inflammatory response.Human EN-RAGE is characterized by two EF-hand calcium-binding motifs, zinc- and copper-binding protein.S100A12 is a disulfide-linked homodimer and the interface between the two subunits is composed mostly of hydrophobic residues.Its proinflammatory activity involves recruitment of leukocytes, promotion of cytokine and chemokine production, and regulation of leukocyte adhesion and migration. EN-RAGE acts as an alarmin or a danger associated molecular pattern (DAMP) molecule and stimulates innate immune cells via binding to receptor for advanced glycation endproducts (AGER). Binding to AGER activates the MAP-kinase and NF-kappa-B signaling pathways leading to production of proinflammatory cytokines and up-regulation of cell adhesion molecules ICAM1 and VCAM1. It also acts as a monocyte and mast cell chemoattractant. Moreover, it can stimulate mast cell degranulation and activation which generates chemokines, histamine and cytokines inducing further leukocyte recruitment to the sites of inflammation. It can inhibit the activity of matrix metalloproteinases; MMP2, MMP3 and MMP9 by chelating Zn2+from their active sites.

  • This recombinant protein can be used for biological assays. For research use only.