Human recombinant protocadherin 10 (from cells)
PRSI91-543
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:PCDH10|KIAA1400|Protocadherin-10
- Protein/peptide name:Protocadherin 10
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:76.38 kD
- Sequence:Gln19-Thr715
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Protocadherin-10 (PCDH10) is a single-pass type I membrane protein that contains six extracellular cadherin domains, one transmembrane domain and one cytoplasmic tail differing from those of the classical cadherins. As a potential calcium-dependent cell-adhesion neuronal receptor, it may plays a role in the establishment and function of specific cell-cell connections in the brain. PCDH10 moderately expressed in all regions of the brain examined, as well as in testis and ovary, and low expression in all other tested tissues.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag