Order Entry
Italy
ContactUsLinkComponent
Human recombinant CD44 (from cells)
Human recombinant CD44 (from cells)
Catalog # PRSI91-517
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CD44 (from cells)
Catalog # PRSI91-517
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Phagocytic Glycoprotein I|MDU2|HUTCH-I|Epican|LHR|CD44|MIC4|ECMR-III|Phagocytic Glycoprotein 1|CDw44|PGP-I|CD44 Antigen|Hyaluronate Receptor|Hermes Antigen|Heparan Sulfate Proteoglycan|Extracellular Matrix Receptor III|MDU3|GP90 Lymphocyte Homing/Adhesion Receptor|PGP-1
  • Protein/peptide name:
    CD44
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.1 kD
  • Sequence:
    Gln21-Pro220
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

CD44 is a cell-surface receptor for hyaluronic acid and also interacts with other ligands, such as osteopontin, collagens, and matrix metalloproteinases. A large number of CD44 isoforms can be generated by the insertion of different combinations of at least nine exons. Increased CD44 antigen is associated with relapses in non-small cell lung cancers. Furthermore, an increasing quantity of evidence suggests that CD44 has various functions related to inflammatory disease. CD44 deficiency induces severe liver injury. CD44-hyaluronate mediates in lymphocyte T and monocyte adhesion to the endothelium, stimulates proinflammatory cytokine release from macrophages and participates in dedifferentiation phenotype of smooth muscle cells from contractile state to synthetic one.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag