Order Entry
Italy
ContactUsLinkComponent
Human recombinant CD300A (from cells)
Human recombinant CD300A (from cells)
Catalog # PRSI91-514
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CD300A (from cells)
Catalog # PRSI91-514
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CMRF35H|CD300a|Immunoglobulin Superfamily Member 12|CMRF35-H9|CD300 antigen-like Family Member A|CLM-8|LMIR1|IRC1/IRC2|CMRF35-H|IgSF12|IRp60|CMRF-35-H9|Inhibitory Receptor Protein 60|NK Inhibitory Receptor|CMRF35-like Molecule 8
  • Protein/peptide name:
    CD300A
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.48 kD
  • Sequence:
    Leu18-Gln178
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

CD300A is a single-pass type I membrane protein that belongs to the CD300 family. CD300A consists of a 163 amino acid (aa) extracellular domain (ECD) with one Ig-like V- type domain, a 21 amino acid transmembrane segment, and a 98 amino acid cytoplasmic domain with tyrosine residues. CD300A is expressed not only by natural killer (NK) cells but also by T-cell subsets, B-cells, dendritic cells, mast cells, granulocytes and monocytes. CD300A is an inhibitory receptor which may contribute to the down-regulation of cytolytic activity in natural killer (NK) cells, and to the down-regulation of mast cell degranulation.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag