Order Entry
Italy
ContactUsLinkComponent
Human Recombinant CCL3L1 (from Cells)
Human Recombinant CCL3L1 (from Cells)
Catalog # PRSI91-466
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant CCL3L1 (from Cells)
Catalog # PRSI91-466
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    G0S19-2|LD78-Beta(1-70)|CCL3L1|G0/G1 Switch Regulatory Protein 19-2|CCL3L3|Tonsillar Lymphocyte LD78 Beta Protein|D17S1718|C-C Motif Chemokine 3-like 1|Small-Inducible Cytokine A3-like 1|SCYA3L1|PAT 464.2
  • Protein/peptide name:
    CCL3L1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    8.82 kD
  • Sequence:
    Ala24-Ala93
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

C-C Motif Chemokine 3-Like 1 (CCL3L1) is a secreted protein that belongs to the intercrine beta (chemokine CC) family. CCL3L1 is a ligand for CCR1, CCR3 and CCR5. CCL3L1 binds to several chemokine receptors including chemokine binding protein 2 and chemokine (C-C motif) receptor 5 (CCR5). CCR5 is a co-receptor for HIV, and binding of this protein to CCR5 inhibits HIV entry. The processed form LD78-beta (3-70) shows a 20-fold to 30-fold higher chemotactic activity and is a very potent inhibitor of HIV-1-infection. The copy number of this gene varies among individuals: most individuals have 1-6 copies in the diploid genome, although rare individuals have zero or more than six copies. The human genome reference assembly contains two full copies of the gene (CCL3L3 and CCL3L1) and a partial pseudogene. This record represents the more centromeric full-length gene.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag